Costar™ Disposable Aspirating Pipets
Costar™ Disposable Aspirating Pipets
Actual product may vary
Costar™ Disposable Aspirating Pipets
Constructed of polystyrene for strength, clarity, and durability. Corning™ Costar™ Disposable Aspirating Pipets are ideal for aseptically aspirating liquids by using vacuum suction.
Have Questions?
Catalog NumberVolume (Metric)Packaging Type
FIS07-200-192 mLIndividual Clear Plastic Wrap
Catalog number FIS07-200-19
Price (SGD)
2,502.00
CS
Add to cart
Volume (Metric):
2 mL
Packaging Type:
Individual Clear Plastic Wrap
Price (SGD)
2,502.00
CS
Add to cart
  • One piece polystyrene construction
  • Sterile and certified nonpyrogenic
  • No graduations; unplugged
  • Tip shape: round
  • Individually wrapped
Specifications
DescriptionAspirating
MaterialPolystyrene
Length (English)10.9 in.
Tip StyleRound
Packaging TypeIndividual Clear Plastic Wrap
Volume (English)0.067 oz.
Volume (Metric)2 mL
Length (Metric)27.7 cm