Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See All Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Boxes
▸
Cryogenic and Freezer Boxes
▸
81- Position Polypropylene Cryo Storage Box
Actual product may vary
Greiner Bio-One
81- Position Polypropylene Cryo Storage Box
Suitable for ultra-low temperature storage of cryogenic vials.
Have Questions?
Catalog Number
Color
FIS07-000-935
Yellow
Catalog number
FIS07-000-935
Price (SGD)
479.00
Case
Add to cart
Color:
Yellow
Price (SGD)
479.00
Case
Add to cart
Holds 1 and 2mL vials
Specifications
Description
Cryo rack with lid, Non sterile
Color
Yellow
For Use With (Application)
Plant tissue culture, transport
Holds
1 and 2 mL Cryo.s™ tubes
Includes
Lid (Natural)
Tube Array
9 x 9