Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Dishes
▸
Petri Dishes
▸
Square Petri Dishes with Vents
Actual product may vary
Greiner Bio-One
Square Petri Dishes with Vents
Manufactured from crystal clear polystyrene.
Have Questions?
Catalog Number
Sterility
FIS07-000-330
Sterile
Catalog number
FIS07-000-330
Price (SGD)
1,148.00
Case
Add to cart
Sterility:
Sterile
Price (SGD)
1,148.00
Case
Add to cart
Clear polystyrene lid
Available sterile or nonsterile
Specifications
Closure Included
Closure Included
Color
Clear
Disposable
Single-use
Material
Polystyrene
Shape
Square
Sterility
Sterile
Type
Petri Dish
Vent
Yes
Height (Metric)
17 mm
Diameter (Metric)
120 mm