Conical Bottom Polystyrene Tubes with Screw Cap
Actual product may vary
Conical Bottom Polystyrene Tubes with Screw Cap
Conical Bottom Polystyrene Tubes with Screw Cap
Greiner Bio-One
Conical Bottom Polystyrene Tubes with Screw Cap
For use in centrifugation and applications that require chemical and thermo resistant vessels.
Have Questions?
Catalog NumberNo. per Pack
FIS07-000-24250
Catalog number FIS07-000-242
Price (SGD)
1,003.00
Case
No. per Pack:
50
Price (SGD)
1,003.00
Case
  • Polystyrene is ideally suited for optical measurements as a result of its high clarity
  • Volume: 15mL
  • Sterile
  • Blue screw cap
  • With graduations and ID writing area
  • Dia. x Height: 17 x 120mm
Specifications
Closure ColorBlue
ColorClear
Capacity (Metric)15 mL
MaterialPolystyrene
No. per Case500
No. per Pack50
SterilitySterile
Bottom ShapeConical
Closure TypeScrew Cap
Closure IncludedYes
Writing AreaYes
GraduatedYes
Product TypeCentrifuge Tube
Unit SizeCase