Pipette Stand
Pipette Stand
Actual product may vary
Pipette Stand
Pipette Stand
Pipette Stand
Accommodate most major brands of pipetters.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPositionsMaterialFor Use With (Equipment)
FIS06-667-26HDPESingle-Channel Pipette
FIS06-666-566AcrylicSingle-Channel Pipette
FIS06-667-43AcrylicSingle-Channel Pipette
Catalog number FIS06-667-2
Price (SGD)
210.00
EA
Add to cart
Positions:
6
Material:
HDPE
For Use With (Equipment):
Single-Channel Pipette
Price (SGD)
210.00
EA
Add to cart
Accommodate most major brands of pipetters.
  • One-position stand (06-667-3) has a single insert hole 1.25 in. (31.8mm) in diameter
  • Three-, four-, and six-position stands have both upper (1.03 in. [26.1mm]) and lower slots (0.625 in. [16mm] in diameter)
  • Multichannel stand is clear acrylic with slots 1 in. (25.5mm) in diameter
Specifications
Product TypePipet Stand
AutoclavableNot Autoclavable
ColorClear, Transparent
HoldsPipettors (6)
MaterialHDPE
Positions6
For Use With (Equipment)Single-Channel Pipette
Dimensions (L x W x H)15.2 x 14 x 22.8 cm (6 x 5.5 x 9 in.)
Unit SizeEA