Blood Collection Tube Dispenser
Blood Collection Tube Dispenser
Actual product may vary
Mitchell Plastics
Blood Collection Tube Dispenser
Have Questions?
Catalog NumberSize
FIS06-666-87
Catalog number FIS06-666-87
Price (SGD)
-
Specifications
For Use With (Application)For Blood collection tubes
For Use With (Equipment)Blood collection tubes
Holds24 Tubes
IncludesTube rack on lid
Length (English) Tube3.93 in.
Length (Metric) Tube100 mm
No. of Places24.0
Product TypeDispenser
Unit SizeEach