▸
Facility Safety and Maintenance
▸
Cleaning Supplies and Equipment
▸
Wipes and Wipe Accessories
▸
Wipe Dispensers
▸
Acrylic Dispenser for Wipes
Actual product may vary
Acrylic Dispenser for Wipes
Acrylic clear holder adds extra weight for one-hand dispensing of tissue wipes.
Have Questions?
Catalog Number
Size
FIS06-666-51
Large
Catalog number
FIS06-666-51
Price (SGD)
91.00
Each
Add to cart
Size:
Large
Price (SGD)
91.00
Each
Add to cart
Acrylic clear holder adds extra weight for one-hand dispensing of tissue wipes.
For use with Kimwipes™
Extra weight for one-hand dispensing
Clear holder mounts to wall or bench with two strips of double-sided tape or mounting screws (not included)
Sized for small or large Kimwipes boxes
Specifications
Product Type
Wiper Dispenser
Color
Clear
Material
Acrylic
Mounting
Free-standing or Wall Mount
For Use With (Equipment)
For Kimwipes™
Size
Large