▸
Facility Safety and Maintenance
▸
Cleaning Supplies and Equipment
▸
Paper Products and Accessories
▸
Paper Products
▸
Scott™ and Kleenex™ Folded Paper Towels
Actual product may vary
Ansell
Scott™ and Kleenex™ Folded Paper Towels
Offer excellent absorbency and hand-drying performance.
Have Questions?
Change view
Catalog Number
Dimensions (L x W)
For Use With (Equipment)
Packaging Quantity
FIS06-666-32B
13.15 x 10.12 in.
C-fold Dispenser
200 Towels/Clip
FIS19-120-476
9.4 x 9.2 in.
Multi-fold towel dispenser
250 Towels/Clip
2 Options
Catalog number
FIS06-666-32B
Price (SGD)
220.00
Case
Add to cart
Dimensions (L x W):
13.15 x 10.12 in.
For Use With (Equipment):
C-fold Dispenser
Packaging Quantity:
200 Towels/Clip
Price (SGD)
220.00
Case
Add to cart
Special drying process produces a softer finished towel
Reliable quality, softness and absorbency means fewer towels used for better economy
Available in C-fold and multifold styles
Specifications
Product Type
C-fold Towel
Length (English)
13.15 in.
Length (Metric)
33.4 cm
Width (English)
13.13 in.
Width (Metric)
25.7 cm
Material
Paper
Certifications/Compliance
EPA, FSC™, EcoLogo™
Color
White
Dimensions (L x W)
13.15 x 10.12 in.
Format
C-Fold
For Use With (Application)
Designed to reduce waste and provides excellent absorbency and hand-drying performance
For Use With (Equipment)
C-fold Dispenser
Packaging Quantity
200 Towels/Clip