Wheaton™ Crimpers
Wheaton™ Crimpers
Actual product may vary
DWK Life Sciences
Wheaton™ Crimpers
With ergonomically designed handles for sealing standard laboratory vials
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
FIS06-451-45920 mm Seals
FIS06-451-45813 mm Seals
Catalog number FIS06-451-459
Price (SGD)
1,400.00
CS
Add to cart
For Use With (Equipment):
20 mm Seals
Price (SGD)
1,400.00
CS
Add to cart
  • Wheaton E-Z Crimpers feature cushioned ergonomic handles that aid in reducing hand fatigue and add a high degree of comfort for the user. Each crimper size is labeled for quick identification. Other features include polished crimping jaws for consistent sealing, a short handle and redesigned spring assembly that further aids in reducing hand fatigue. For semi-automatic crimping, use the E-Z Crimper with the Wheaton Crimpmaster™ Crimping Station. An adjustable stop allows you to determine the tightness of the seal while maintaining consistent crimping. Tapered jaws provide for easier vial access when using racks. The 11 mm E- Z Crimper is supplied with a plunger insert to give the option of crimping the top of a vial creating an extra tight seal. We recommend this only for standard opening vials.
  • Crimpers can be autoclaved, although autoclaving may change the appearance or reduce product life. They should not be autoclaved at greater than 121°C
Specifications
QuantityCase of 1
For Use With (Equipment)20 mm Seals
Unit SizeCS