Check Order Status
Wheaton™ Decappers
Actual product may vary
DWK Life Sciences
Wheaton™ Decappers
These crimpers are used for flip cap seals and for aluminum seals
Have Questions?
Catalog Number
Closure Type
Type
FIS06-451-136
20 mm Caps
Plier-style Decapper
Catalog number
FIS06-451-136
Price (SGD)
424.00
EA
Add to cart
Closure Type:
20 mm Caps
Type:
Plier-style Decapper
Price (SGD)
424.00
EA
Add to cart
Specifications
Closure Type
20 mm Caps
Type
Plier-style Decapper
For Use With (Equipment)
Flip Cap Seals and Aluminum Seals