▸
Vials
▸
Autosampler Vials, Inserts, and Closures
▸
Chromatography Autosampler Vial Caps Only
▸
Wheaton™ Unlined Aluminum Open-Top Seals
Actual product may vary
DWK Life Sciences
Wheaton™ Unlined Aluminum Open-Top Seals
Open-top seal exposes stopper for sample retrieval with a syringe
Have Questions?
Catalog Number
Color
Closure Size
FIS06-449-21
Natural
13 mm
Catalog number
FIS06-449-21
Price (SGD)
318.00
Case
Add to cart
Color:
Natural
Closure Size:
13 mm
Price (SGD)
318.00
Case
Add to cart
Unlined
Closure Type: Seal
Specifications
Color
Natural
Material
Aluminum
Quantity
1000/Cs.
Closure Size
13 mm
Product Type
Open Top Cap