Wheaton™ Stoppers for Serum Bottles and Vials: 3-Leg Lyophilization Stopper
Wheaton™ Stoppers for Serum Bottles and Vials: 3-Leg Lyophilization Stopper
Actual product may vary
DWK Life Sciences
Wheaton™ Stoppers for Serum Bottles and Vials: 3-Leg Lyophilization Stopper
Provide airtight covers
Have Questions?
Catalog NumberColor
FIS06-447HGray
Catalog number FIS06-447H
Price (SGD)
809.00
CS
Add to cart
Color:
Gray
Price (SGD)
809.00
CS
Add to cart
Provide airtight covers
  • Flange-type stoppers are secured with aluminum crimp seals
  • Slotted and three-prong plug stoppers are for freeze-dry applications
Specifications
Diameter (Metric) Inner2 cm
For Use With (Application)Lyophilization
For Use With (Equipment)Fits 1.3cm I.D., 2cm O.D. mouth of serum bottles and vials
Hardness50 Durometer
AutoclavableAutoclavable
ColorGray
MaterialButyl Rubber
Product Type3-Leg Lyophilization Stopper
Unit SizeCS