Check Order Status
Wheaton™ Stoppers for Serum Bottles and Vials: 2-Leg Stoppers
Actual product may vary
DWK Life Sciences
Wheaton™ Stoppers for Serum Bottles and Vials: 2-Leg Stoppers
Provide airtight covers
Have Questions?
Catalog Number
Product Type
Material
FIS06-447E
2-Leg Lyophilization Stopper
Butyl Rubber
Catalog number
FIS06-447E
Price (SGD)
548.00
CS
Add to cart
Product Type:
2-Leg Lyophilization Stopper
Material:
Butyl Rubber
Price (SGD)
548.00
CS
Add to cart
Provide airtight covers
Flange-type, straight plug stoppers are secured with aluminum crimp seals (06-406-11B)
Three-prong plug stoppers are for freeze-dry applications (06-447E, 06-451-56)
Specifications
Diameter (Metric)
13 mm
Diameter (Metric) Inner
7 mm
For Use With (Application)
Lyophilization
For Use With (Equipment)
Fits 7mm I.D., 13mm O.D. mouth of serum bottles and vials
Hardness
50 Durometer
Autoclavable
Autoclavable
Color
Gray
Material
Butyl Rubber
Product Type
2-Leg Lyophilization Stopper