Wheaton™ Serum Vials
Wheaton™ Serum Vials
Actual product may vary
Wheaton™ Serum Vials
Wheaton™ Serum Vials
DWK Life Sciences
Wheaton™ Serum Vials
Available as clear and amber vials
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)ColorDiameter (Metric) Outer
FIS06-446E20 mLClear30.5 mm
FIS06-406-372 mLClear15 mm
FIS06-446C3 mLClear17 mm
FIS06-446D5 mLClear22 mm
Catalog number FIS06-446E
Price (SGD)
429.00
CS
Add to cart
Volume (Metric):
20 mL
Color:
Clear
Diameter (Metric) Outer:
30.5 mm
Price (SGD)
429.00
CS
Add to cart
Available as clear and amber vials
Glass can withstand the challenges of reduced pressure and extreme temperature variations. Protects against changes in pH and maintain the purity of the contents.
  • Clear vials made from low extractable borosilicate glass tubing to ensure uniform wall thickness
  • Amber vials made from borosilicate glass conforms to USP Type 1 requirements to protect light-sensitive samples
  • Withstand extreme temperature variations
  • Necks finished for aluminum crimp seals
  • Blowback feature of neck and specially designed bottom radius for lyophilization applications
  • Packaged in shrink-wrapped modules to reduce particulate contamination
Lyophilization, packaging, and sampling
Specifications
Diameter (English) Inner0.51 in.
Diameter (Metric) Inner13 mm
ColorClear
Diameter (Metric) Outer30.5 mm
MaterialBorosilicate Glass
Volume (Metric)20 mL
For Use With (Application)For lyophilization, packaging, sampling and freeze drying applications
Volume (English)5 dr.
Product LineWheaton
Volume (Metric) Total20 mL
AutoclavableAutoclavable
Closure TypeCrimp Cap
Height (Metric)58 mm
Certifications/ComplianceASTM E438, USP Type I, Type I, Class A
TypeSerum Vial
Unit SizeCS