Wheaton™ Dounce Tissue Grinders
Wheaton™ Dounce Tissue Grinders
Actual product may vary
DWK Life Sciences
Wheaton™ Dounce Tissue Grinders
Ideal for enzyme studies. Wheaton™ Dounce Tissue Grinders are designed to retain a high percentage of cell nuclei, mitochondria, and proteins in soft tissues or cell suspensions.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacityLength (English)Length (English) TubeTube Diameter (English) Outside
FIS06-435C1.35 oz., 40mL11.2 in.5.5 in.1.25 in.
FIS06-435B0.5 oz., 15mL8.26 in.3.7 in.0.86 in.
FIS06-4340.03 oz., 1mL4.92 in.1.8 in.0.43 in.
Catalog number FIS06-435C
Price (SGD)
686.00
EA
Add to cart
Capacity:
1.35 oz., 40mL
Length (English):
11.2 in.
Length (English) Tube:
5.5 in.
Tube Diameter (English) Outside:
1.25 in.
Price (SGD)
686.00
EA
Add to cart
Ideal for enzyme studies. Wheaton™ Dounce Tissue Grinders are designed to retain a high percentage of cell nuclei, mitochondria, and proteins in soft tissues or cell suspensions.
  • Sample covers the pestle ball, preventing heat buildup by reducing friction against the mortar
  • Mortar features large reservoir and pouring lip
  • Supplied with “loose” and “tight” pestles
  • Components are made of low extractable borosilicate glass
Specifications
Capacity (English)1.35 oz.
Capacity (Metric)40 mL
MaterialBorosilicate Glass
Product TypeDounce Tissue Grinder
For Use With (Application)Designed to retain a high percentage of cell nuclei, mitochondria and Proteins in soft tissues or cell suspensions
Length (English)11.2 in.
Length (Metric)285mm
Length (English) Tube5.5 in.
Length (Metric) Tube140mm
Tube Diameter (English) Outside1.25 in.
Tube Diameter (Metric) Outside32mm
Capacity1.35 oz., 40mL
Unit SizeEA