Check Order Status
NFκB p52 Rabbit anti-Human, Polyclonal, MilliporeSigma™
Actual product may vary
NFκB p52 Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Have Questions?
Catalog number
FIS06-413
Price (SGD)
1,276.00
CS
Add to cart
Price (SGD)
1,276.00
CS
Add to cart
Detect NFκB p52 with Anti-NFκB p52 Antibody (Rabbit Polyclonal Antibody), that has been shown to work in immunoprecipitation and western blotting.
For Research Use Only
Specifications
Antigen
NFκB p52
Applications
Immunoprecipitation, Western Blot
Classification
Polyclonal
Conjugate
Unconjugated
Formulation
Protein A Purified immunoglobulin in in 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% NaN3 as a preservative.
Gene Symbols
NFKB2; LYT10; LYT-10; H2TF1; Lyt10
Host Species
Rabbit
Immunogen
KLH conjugated synthetic peptide (CYNPGLDGIIEYDDFK) contained within amino acids 4-19 of human NF κB, p52
Purification Method
Unpurified
Quantity
100 μL
Primary or Secondary
Primary
Gene ID (Entrez)
NM_019408
Target Species
Human
Form
Purified
Isotype
IgG
Unit Size
CS
Contents & Storage
−20°C in undiluted aliquotes for 24 months. Avoid repeated freeze/thaw cycles.