Wheaton™ Serum Bottles and Vials
Actual product may vary
Wheaton™ Serum Bottles and Vials
Wheaton™ Serum Bottles and Vials
DWK Life Sciences
Wheaton™ Serum Bottles and Vials
These bottles have exceptional wall strength and superior heat and chemical tolerances
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Color
FIS06-406G50 mLClear
FIS06-406C5 mLClear
FIS06-402F100 mLAmber
FKS10011422100 mLAmber
FIS06-406J100 mLClear
FIS06-406K125 mLClear
Catalog number FIS06-406G
Price (SGD)
1,684.00
Case
Capacity (Metric):
50 mL
Color:
Clear
Price (SGD)
1,684.00
Case
These bottles have exceptional wall strength and superior heat and chemical tolerances
Ideal for long- and short-term sample storage and vaccine/injectable drug containers.
  • Clear borosilicate glass conforms to USP Type 1 requirements
  • Amber borosilicate glass conforms to USP Type 1 requirements
  • Shrink wrapped partitioned modules reduce breakage
Specifications
Capacity1.7 oz., 50 mL
Capacity (Metric)50 mL
Capacity (English)1.7 oz.
ColorClear
Diameter (English)1.69 in.
MaterialBorosilicate Glass
Height (Metric)73 mm
Height (English)2.87 in.
Diameter (Metric)43 mm
Unit SizeCase