▸
Tubes
▸
Tube Caps and Closures
▸
Morton™ Stainless-Steel Culture Tube Closures
Actual product may vary
Bal Supply
Morton™ Stainless-Steel Culture Tube Closures
Ideal for use with standard glass culture tubes
Have Questions?
Change view
Catalog Number
Product Type
Size (Metric)
FIS05-888A
Lanced Closure
13 mm
FIS05-888-1C
Culture Tube Closure
18 mm
FIS05-888-1A
Culture Tube Closure
13 mm
FIS05-888-1B
Culture Tube Closure
16 mm
4 Options
Catalog number
FIS05-888A
Price (SGD)
-
Product Type:
Lanced Closure
Size (Metric):
13 mm
Ideal for use with standard glass culture tubes
Non-contaminating, noncorrosive stainless-steel cylindrical closures
Top surface indentations allow adequate gas exchange under aerobic and anaerobic conditions, yet prevent buildup of dust and microorganisms
Reduce amount of specimen evaporation by 50% as compared to cotton plugging
Withstand repeated autoclaving without loss of resiliency
Available as plain or lanced closures
Lanced closures feature three resilient fingers that adjust to nominal variations in tube O.D
Specifications
Product Type
Lanced Closure
Autoclavable
Autoclavable
Material
Stainless Steel
Size (English)
1/2 in.
Size (Metric)
13 mm
For Use With (Equipment)
Standard glass culture tubes for microbiology and related applications