Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Tubes
▸
Tube Caps and Closures
▸
Morton™ Stainless-Steel Culture Tube Closures
Actual product may vary
Bal Supply
Morton™ Stainless-Steel Culture Tube Closures
Ideal for use with standard glass culture tubes
Have Questions?
Change view
Catalog Number
Product Type
Size (Metric)
FIS05-888-1A
Culture Tube Closure
13 mm
FIS05-888-1C
Culture Tube Closure
18 mm
FIS05-888-1B
Culture Tube Closure
16 mm
FIS05-888A
Lanced Closure
13 mm
4 Options
Catalog number
FIS05-888-1A
Price (SGD)
669.00
Pack
Add to cart
Product Type:
Culture Tube Closure
Size (Metric):
13 mm
Price (SGD)
669.00
Pack
Add to cart
Ideal for use with standard glass culture tubes
Non-contaminating, noncorrosive stainless-steel cylindrical closures
Top surface indentations allow adequate gas exchange under aerobic and anaerobic conditions, yet prevent buildup of dust and microorganisms
Reduce amount of specimen evaporation by 50% as compared to cotton plugging
Withstand repeated autoclaving without loss of resiliency
Available as plain or lanced closures
Lanced closures feature three resilient fingers that adjust to nominal variations in tube O.D
Specifications
Product Type
Culture Tube Closure
Autoclavable
Autoclavable
Material
Stainless Steel
Size (English)
1/2 in.
Size (Metric)
13 mm
For Use With (Equipment)
Standard glass culture tubes for microbiology and related applications