Hoffman Tubing Clamps
Hoffman Tubing Clamps
Actual product may vary
Humboldt
Hoffman Tubing Clamps
Regulate flow from fully open to closed
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHeight (English)Height (Metric)Jaw TypeSize
FIS05-870B0 to 0.75 in.0 to 19 mmFixed1 in.
FIS05-871B0 to 0.75 in.0 to 19 mmSwivel1 in.
FIS05-871A0 to 0.62 in.0 to 16 mmSwivel0.75 in.
Catalog number FIS05-870B
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Height (English):
0 to 0.75 in.
Height (Metric):
0 to 19 mm
Jaw Type:
Fixed
Size:
1 in.
Regulate flow from fully open to closed
  • For flexible tubing
  • Nickel-plated brass
  • Won't rust
  • Lower jaw is either fixed or swivel-mounted for easy insertion of tubing without disconnecting apparatus
Specifications
Quantity1 Ea.
TypeTubing Clamp
MaterialNickel-plated Brass
For Use With (Equipment)Control The Flow in Flexible Tubing
For Use With (Application)Control the flow in flexible tubing
Height (Metric)0 to 19 mm
Height (English)0 to 0.75 in.
Size1 in.
Jaw TypeFixed
Unit SizeEA