Round Covered Lead Flask Rings
Round Covered Lead Flask Rings
Actual product may vary
Fisherbrand
Round Covered Lead Flask Rings
Stabilize Erlenmeyer flasks immersed in water baths.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (English) InnerDiameter (Metric) InnerWeight (English)Weight (Metric)
FIS05-86982.7 in.7 cm2 lb.907.1 g
FIS05-869-62 in.5.1 cm1 lb.453.5 g
FIS05-86962 in.5.1 cm1 lb.453.5 g
FIS05-86951.6 in.4.1 cm0.5 lb.226.7 g
FIS05-869-72.2 in.5.6 cm1.5 lb.680.3 g
FIS05-86972.2 in.5.6 cm1.5 lb.680.3 g
Catalog number FIS05-8698
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Diameter (English) Inner:
2.7 in.
Diameter (Metric) Inner:
7 cm
Weight (English):
2 lb.
Weight (Metric):
907.1 g
Stabilize Erlenmeyer flasks immersed in water baths.
  • Reduces floating, breakage and chemical spills
  • Bright blue coating
  • Chemical-resistant
  • Will not scratch flasks
  • Available in four diameter/weight combinations
Specifications
Product TypeLead Ring
AutoclavableNot Autoclavable
ColorBlue
HoldsFlasks (1 to 4 L)
MaterialLead
ShapeRound
ShelvesNone
StackableYes
SterilityNon-sterile
For Use With (Equipment)1000 to 4000mL Erlenmeyer flasks immersed in water baths
Length (Metric)114 mm
Width (Metric)43 mm
Height (Metric)114 mm
Diameter (English) Inner2.7 in.
Diameter (Metric) Inner7 cm
Weight (English)2 lb.
Weight (Metric)907.1 g
Unit SizeEA