Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Clamps and Supports
▸
Clamps
▸
Day Pinchcock
Actual product may vary
Humboldt
Day Pinchcock
Can be put on or removed without disconnecting apparatus
Have Questions?
Catalog Number
Length (Metric)
FIS05-867
6.65 cm
Catalog number
FIS05-867
Price (SGD)
7.00
Each
Add to cart
Length (Metric):
6.65 cm
Price (SGD)
7.00
Each
Add to cart
Can be put on or removed without disconnecting apparatus
Tough 302 Stainless Steel
Holds flexible tubing up to 0.37 in. (9.5mm) O.D.
Specifications
Quantity
1 Ea.
Type
Pinchcock Clamp
Material
Stainless Steel (302)
Length (English)
2.62 in.
Length (Metric)
6.65 cm