Castaloy™ Hosecock Clamp
Castaloy™ Hosecock Clamp
Actual product may vary
Fisherbrand
Castaloy™ Hosecock Clamp
Fine-machined adjustment screw for very accurate regulation of flow
Have Questions?
Catalog NumberFor Use With (Equipment)
FIS05-847QAccurate Regulation of Flow
Catalog number FIS05-847Q
Price (SGD)
-
For Use With (Equipment):
Accurate Regulation of Flow
Fine-machined adjustment screw for very accurate regulation of flow
  • Threaded keeper screw on side of clamp can be raised to allow easy positioning of tubing, then screwed back into base of clamp
  • For tubing up to 0.75 in. (19mm) O.D.
  • Concave lower bearing surface, convex upper which reduces amount of pressure required for complete closure
Specifications
Quantity1 Ea.
TypeHosecock Clamp
IncludesAdjustment screw
For Use With (Equipment)Accurate Regulation of Flow
For Use With (Application)For accurate regulation of flow
Size (Metric) Grip19 mm O.D.
Size (English) Grip0.75 in. O.D.
Unit SizeEA