Nalgene™ Polypropylene Pinch Clamp
Nalgene™ Polypropylene Pinch Clamp
Actual product may vary
Thermo Scientific
Nalgene™ Polypropylene Pinch Clamp
From completely open to restricted flow to completely closed—experience precise, one-hand regulation of fluid flow with the Thermo Scientific™ Nalgene™ Polypropylene Pinch Clamp.
Have Questions?
Catalog NumberQuantity
FIS05-835BCase of 72
Catalog number FIS05-835B
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
Case of 72
From completely open to restricted flow to completely closed—experience precise, one-hand regulation of fluid flow with the Thermo Scientific™ Nalgene™ Polypropylene Pinch Clamp. This plastic clamp will not rust, corrode or deform. Molded in one piece with no sharp edges, it accommodates a wide range of tubing.
Specifications
QuantityCase of 72
TypePinch Clamp
MaterialPolypropylene
For Use With (Equipment)Fits tubing with O.D.: 1/4 to 7/16 in.
DescriptionPinch clamp; Fits tubing with O.D.: 1/4 to 7/16 in.
Unit SizePK