Check Order Status
Nylon Screw Compressor Clamp
Actual product may vary
Bel-Art
Nylon Screw Compressor Clamp
Tubing clamp with angled jaws prevents tube from slipping out
Have Questions?
Catalog Number
Color
FIS05-834
Brown
Catalog number
FIS05-834
Price (SGD)
168.00
PK
Add to cart
Color:
Brown
Price (SGD)
168.00
PK
Add to cart
Tubing clamp with angled jaws prevents tube from slipping out
Open-jaw nylon tubing clamp fits any tubing up to 1/4 in. O.D. (6.4mm)
Thumbscrew clamp applies even force
Specifications
Quantity
12 Pk.
Type
Screw Compressor Clamp
Material
Nylon
For Use With (Equipment)
0.25 in. O.D. Tube
Length (English)
2.31 in.
Length (English) Opening
0 to 0.25 in.
Length (Metric)
5.867 cm
Length (Metric) Opening
0 to 0.63 cm
Color
Brown
Jaw Type
Angled
Diameter (Metric) Inside
6.4 mm