Check Order Status
Castaloy™ Clamp Jumbo Holder
Actual product may vary
Castaloy™ Clamp Jumbo Holder
Safe for heavy loads
Have Questions?
Catalog number
FIS05-757Q
Price (SGD)
-
Safe for heavy loads
Grips in variety of positions
Grips rods up to 0.87 in. (22mm) diameter
Holds flat shelf or panel to 0.75 in. (19mm) thick
Made of diecast aluminum
With 45° angle thumbscrews
Specifications
Quantity
1 Ea.
Type
Jumbo Clamp Holder
Material
Die Cast Aluminum
Includes
45° Angle thumbscrews