PYREX™ Chromatography Column With Coarse Fritted Disc
PYREX™ Chromatography Column With Coarse Fritted Disc
Actual product may vary
PYREX™ Chromatography Column With Coarse Fritted Disc
Designed for general chromatographic analysis
Have Questions?
Catalog NumberLength (Metric)Diameter (Metric) Inner
FIS05-722B400 mm22 mm
Catalog number FIS05-722B
Price (SGD)
1,964.00
Case
Add to cart
Length (Metric):
400 mm
Diameter (Metric) Inner:
22 mm
Price (SGD)
1,964.00
Case
Add to cart
These tubes are designed for general chromatographic analysis. The tops are tooled for rubber stopper fit and the tubes are fabricated with sealed-in coarse porosity fritted discs. The outflow tubes are large in diameter to eliminate the smearing of the definition bands of effluent caused by the tube being filled, and the solvent mixed before being expelled. The outflow tubes are long enough to protrude through a rubber stopper.
Specifications
For Use With (Application)General chromatographic analysis
Diameter (Metric) Inner22 mm
TypeChromatography Column
Pore Size40 to 60 μm
Column Capacity125 mL
Length (Metric)400 mm
MaterialBorosilicate Glass
IncludesFritted disc
Stopper Number4
Unit SizeCase