UV Darkroom Cabinet
Actual product may vary
UV Darkroom Cabinet
UV Darkroom Cabinet
Spectroline
UV Darkroom Cabinet
Portable, space-saving mini-cabinet doubles as a personal darkroom and UV irradiating units
Have Questions?
Catalog NumberDimensions (L x W x H)
FIS05-720-1012 x 9 x 6.5 in.
Catalog number FIS05-720-10
Price (SGD)
2,309.00
Each
Dimensions (L x W x H):
12 x 9 x 6.5 in.
Price (SGD)
2,309.00
Each
Portable, space-saving mini-cabinet doubles as a personal darkroom and UV irradiating units
  • Flexible, contoured eyepiece with a built-in UV-absorbing window for safe sample viewing
  • Felt curtain permits easy access to cabinet interior while shielding samples from external light
  • Removable bottom panel allows easy positioning over transilluminators
  • Lightweight, durable construction creates easily transportable darkroom
  • Wavelength: UV (254nm-365nm)
  • Wattage: 6w
  • LxWxH: 30.5x16.5cm (12x6.5 in.)
Specifications
Dimensions (L x W x H)12 x 9 x 6.5 in.
Height (English)6.5 in.
Height (Metric)165.1 mm
Length (English)12 in.
Length (Metric)304.8 mm
Packaging Quantity1 Ea.
Width (English)9 in.
Width (Metric)228.6 mm
Product TypeUV Darkroom Cabinet
Unit SizeEach