DURAN™ Screw Cap GL 45 with 2-Hose Connector
Actual product may vary
DURAN™ Screw Cap GL 45 with 2-Hose Connector
DURAN™ Screw Cap GL 45 with 2-Hose Connector
DWK Life Sciences
DURAN™ Screw Cap GL 45 with 2-Hose Connector
The DURAN™ Screw Cap GL 45 with 2-Hose Connector provides a versatile, reusable two-port connection system for DURAN™ GL 45 bottles. Ideal for liquid transfer via peristaltic pump, it supports silicone tubing with 6 to 9 mm inner diameter and is fully autoclavable.
Have Questions?
Catalog NumberThread Size (Metric)
FIS05-719-310GL 45
Catalog number FIS05-719-310
Price (SGD)
412.00
Pack
Thread Size (Metric):
GL 45
Price (SGD)
412.00
Pack
  • DURAN™ GL 45 screw cap with two hose connectors for easy connection of flexible tubing to GL 45 laboratory and media bottles
  • Connectors suitable for soft elastic silicone tubing with an inner diameter of 6 to 9 mm
  • Grey polypropylene cap centre rotates freely, allowing the bottle to be unscrewed without disconnecting the tubing
  • EPDM gasket ensures an airtight, liquid-tight seal with the bottle
  • Temperature resistant up to +140°C; autoclavable at 121°C and dishwasher-proof
  • Based on the standard GL 45 thread, highly versatile and compatible across DURAN™ GL 45 bottle range
  • Optional venting connector available with or without a syringe filter (0.2 μm)
  • Typical applications include transfer of sterile media between containers using a peristaltic pump in biotech and life science workflows
Specifications
DescriptionDURAN Screw Cap GL 45 with 2 hose connections EDPM Gasket
For Use With (Application)Sterile fluid transfer in and out of lab bottles; Gas transfer in and out of lab bottles; Fluid transfer bewteen lab bottles; Supply of reagents to instruments
MaterialPolypropylene, EPDM
Quantity2/Pk.
Thread Size (Metric)GL 45
Neck Finish93 mm