Wide-Mouth Tall-Profile Clear Glass Jars with Closure
Actual product may vary
Wide-Mouth Tall-Profile Clear Glass Jars with Closure
Wide-Mouth Tall-Profile Clear Glass Jars with Closure
Wide-Mouth Tall-Profile Clear Glass Jars with Closure
Thermo Scientific
Wide-Mouth Tall-Profile Clear Glass Jars with Closure
Collect soil, sludge, waste and water samples for a variety of organic and inorganic target analytes with these wide-mouth, tall-profile clear glass jars with closure when light protection is not required.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)LevelClosure Size
FIS05-719-281A2000 mLUnprocessed83-400 mm
FIS05-719-68500 mLUnprocessed70-400 mm
FIS05-719-81950 mLUnprocessed89-400 mm
Catalog number FIS05-719-281A
Price (SGD)
-
Capacity (Metric):
2000 mL
Level:
Unprocessed
Closure Size:
83-400 mm
Collect soil, sludge, waste and water samples for a variety of organic and inorganic target analytes with Thermo Scientific™ Wide-Mouth Tall-Profile Clear Glass Jars when light protection is not required. Jars are available Certified to meet EPA performance-based specifications for semi-volatile organics, pesticides, PCBs and metals anlyses, Processed the same way as the certified jars but without certification documentation, or Unprocessed and ready for your own cleaning procedure.
  • Clear Type III soda-lime glass jar
  • PTFE-lined white polypropylene closed-top cap
  • Sizes range from 125 to 4000mL
  • Closures assembled on bottles
Certified jars come with the following features:
  • Certified to meet or exceed EPA performance-based standards for semi-volatile organics, pesticides, PCBs and metals analyses
  • Certificate of Analysis (CofA) included in the case and barcoded with lot number for traceability
  • Each jar individually barcoded with lot number and unique container number
  • Analysis labels included unattached for optional use
  • Carton sealed with security tape providing tamper evidence
  • Chipboard divider packaging to cushion and help prevent breakage; overpack required for shipping
Processed jars are processed in the same manner as certified jars and come with all the features of certified jars, but jars are not barcoded, and CofA is not included.
Unprocessed jars are ready for your own in-house cleaning and certification process. Boxed with chipboard dividers to help prevent breakage; overpack required for shipping.
Specifications
Capacity64 oz., 2000 mL
Capacity (Metric)2000 mL
Capacity (English)64 oz.
Certification ParametersNone
ColorClear
DescriptionClear Glass Tall Jar
Diameter (English)4.88 in.
Diameter (English) Inner Neck2.7 in.
Diameter (Metric) Inner Neck68.5 mm
MaterialGlass (Type 3)
No. per Case6
Replacement ClosureB167-8300BL
Closure ColorWhite
LevelUnprocessed
Liner MaterialPTFE
Volume (English) Brim67.5 oz.
Volume (Metric) Brim1996 mL
Certifications/ComplianceNone
MouthWide
Height (Metric)214 mm
Product TypeJar
Height (English)8.42 in.
Closure MaterialPolypropylene
Closure Size83-400 mm
Diameter (Metric)124 mm
Closure TypeClosed-top Cap
Unit SizeCase