Whatman™ 903 Protein Saver Card
Whatman™ 903 Protein Saver Card
Actual product may vary
Fisherbrand
Whatman™ 903 Protein Saver Card
For research use only
Have Questions?
Catalog number FIS05-715-121
Price (SGD)
824.00
PK
Add to cart
Price (SGD)
824.00
PK
Add to cart
For research use only
  • Each card has five half-inch circles printed on the sample collection area
  • Each circle holds 75 to 80 μL of sample
  • Wraparound cover has spaces for name and date of collection and is imprinted with universal biohazard symbol in accordance with USPS regulations
  • Fits into Whatman foil barrier zipper bags for storage
Specifications
For Use With (Application)Research use only
IncludesFive 0.5 in. circles holding 75 to 80 μL sample, wraparound cover with spaces for name and date of collection, imprinted with universal biohazard symbol
Quantity100/Pk.
TypeProtein Saver Card
Unit SizePK