Lamin A/C Mouse anti-Human, Mouse, Rat, Canine, Chicken, Clone: 14, MilliporeSigma™
Lamin A/C Mouse anti-Human, Mouse, Rat, Canine, Chicken, Clone: 14, MilliporeSigma™
Actual product may vary
Fisher Catalogue
Lamin A/C Mouse anti-Human, Mouse, Rat, Canine, Chicken, Clone: 14, MilliporeSigma™
Mouse Monoclonal Antibody
Have Questions?
Catalog number FIS05-714
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Anti-Lamin A/C Antibody, clone 14 detects level of Lamin A/C and has been published and validated for use in IC and western blotting.
For Research Use Only
Specifications
AntigenLamin A/C
ApplicationsImmunocytochemistry, Western Blot
ClassificationMonoclonal
Clone14
ConjugateUnconjugated
FormulationPurified mouse monoclonal IgG1 in buffer containing 50% storage buffer (20mM sodium phosphate, pH 7.5, 0.15MNaCl, 1mg/mL BSA, 0.09% sodium azide) and 50% glycerol. Store at −20°C.
Gene Accession No.NM_005572
Gene SymbolsLMNA, HGPS, EMD2, FPLD, CDCD1, LDP1, LGMD1B, IDC, CMT2B1, LMNC, FPL, PRO1, Lamin-A/C, LMN1, CMD1A, LFP, CDDC
Host SpeciesMouse
ImmunogenPeptide from human lamin A/C corresponding to amino acids 398 to 490.
Purification MethodProtein G purfied
Quantity50 μg
Primary or SecondaryPrimary
Target SpeciesCanine, Chicken, Human, Mouse, Rat
FormPurified
IsotypeIgG
Unit SizePK
Contents & Storage
−20°C for up to 12 months.