▸
Chromatography Systems
▸
Liquid Chromatography Instruments
▸
Individual HPLC Components and Accessories
▸
HPLC Solvent Handling Component Accessories
▸
Inlet Solvent Filters
Actual product may vary
Idex
Inlet Solvent Filters
Stainless-steel or multiple materials
Have Questions?
Catalog Number
Pore Size
Includes
FIS05-701-444
20 μm
1/pk.
Catalog number
FIS05-701-444
Price (SGD)
-
Pore Size:
20 μm
Includes:
1/pk.
Cups and end fittings sold separately, see 05-704-54 and 045-707-83.
Specifications
Flow Rate
100 mL/min.
Includes
1/pk.
Material
Stainless Steel, PCTFE
Pore Size
20 μm
Thread Size
N/A
Type
Inlet Solvent Filter