FEP Chromatography Tubing: 100 ft.
Actual product may vary
Idex
FEP Chromatography Tubing: 100 ft.
Excellent chemical resistance at an economical price
Have Questions?
Catalog NumberOverall Length (Metric)
FIS05-701-16830.4 m
Catalog number FIS05-701-168
Price (SGD)
-
Overall Length (Metric):
30.4 m
Excellent chemical resistance at an economical price
  • Great for moderate to low pressure applications
  • Low gas permeability
  • Max pressure 500 psi
Specifications
TypeFEP Tubing
Certifications/ComplianceROHS
AutoclavableAutoclavable
ColorNatural
pH Range0 to 14
Tolerance±0.003 in.
Sterilization MethodEthylene Oxide, Thermal
For Use With (Application)Analytical Instrumentation, In Vitro Diagnostics, Biotechnology, Laboratory and General, Low Pressure AppliCatIons
Diameter (English) Inner0.062 in.
Diameter (English) Outer0.12 in.
Diameter (Metric) Inner1.574 mm
Diameter (Metric) Outer0.3 cm
Overall Length (English)100 ft.
Overall Length (Metric)30.4 m
MaterialFEP
Maximum Pressure500 psi
Min. Operating Temperature (Metric)-59.8°F
Max. Operating Temperature (Metric)50°C
Unit SizeEach