PYREX™ Round-Bottom Glass Centrifuge Tubes
PYREX™ Round-Bottom Glass Centrifuge Tubes
Actual product may vary
PYREX™ Round-Bottom Glass Centrifuge Tubes
Without graduations
Have Questions?
Catalog NumberLength (Metric)Closure IncludedClosure Material
FIS05-550116 mmNoN/A
Catalog number FIS05-550
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
116 mm
Closure Included:
No
Closure Material:
N/A
These PYREX™ round bottom, ungraduated centrifuge tubes have a pourout spout for easy decanting.
Specifications
Closure MaterialN/A
Capacity (Metric)50 mL
DisposableReusable
MaterialBorosilicate Glass
For Use WithUseful for the handling of sputum specimens, digestion, shaking, neutralizing and centrifuging
No. per Case12
No. per Pack12
SterilityNon-sterile
ShapeCylindrical
Length (Metric)116 mm
Closure IncludedNo
Diameter (Metric)29 mm
Product TypeCentrifuge Tube
Unit SizeEA