Search Thermo Fisher Scientific
| Catalog Number | Sterility | Capacity (Metric) |
|---|---|---|
| FIS05-541-62 | Non-sterile | 1.5 mL |
| FIS05-541-61 | Sterile | 0.5 mL |
| FIS05-541-63 | Sterile | 1.5 mL |
Extremely low surface adhesion, which is ideal for protein or nucleic acid studies to prevent any possible loss of sample due to adherence to the inner walls of the tube allowing complete sample recovery.