Search Thermo Fisher Scientific
| Catalog Number | Capacity (Metric) | Max. RCF | Closure Type |
|---|---|---|---|
| FIS05-538-68 | 50 mL | 3000 x g | CentriStar |
| FKS10151151 | 15 mL | 12,000 x g | Plug |
| FIS05-538-53D | 15 mL | 12,000 x g | Plug |
| FKS10431013 | 15 mL | 12,000 x g | Plug |
| FIS05-538-55 | 50 mL | 15,500 x g | Plug |
| FKS10314131 | 50 mL | 15,500 x g | Plug |
| FIS05-538-55A | 50 mL | 15,500 x g | Plug |
| FKS10694541 | 50 mL | 15,500 x g | Plug |
| FKS10038980 | 50 mL | 3000 x g | CentriStar |
| FIS05-538-67 | 50 mL | 3000 x g | Plug |
| FKS10509891 | 50 mL | 3000 x g | Plug |
| CRG430776 | 250 mL | 6000 x g | Plug |
| FIS05-538-53 | 250 mL | 6000 x g | Plug |
| FKS10758961 | 250 mL | 6000 x g | Plug |
| FIS07-200-621 | 500 mL | 6000 x g | Plug |
| FKS10018910 | 500 mL | 6000 x g | Plug |