15mL Graduated Polystyrene Centrifuge Tube
Actual product may vary
15mL Graduated Polystyrene Centrifuge Tube
Excellent general-purpose tube
Have Questions?
Catalog NumberCapacity (Metric)
FIS05-527-4515 mL
Catalog number FIS05-527-45
Price (SGD)
559.00
Case
Capacity (Metric):
15 mL
Price (SGD)
559.00
Case
Excellent general-purpose tube
  • With conical bottom
  • Nonsterile polystyrene
  • Features molded-in graduations in 0.1mL increments up to 1mL and in 0.5mL subdivisions between 1 and 15mL
Specifications
Diameter (English) Outside0.7 in.
Capacity (Metric)15 mL
MaterialPolystyrene
No. per Case1000
SterilityNon-sterile
ShapeConical
Length (Metric)120 mm
Capacity (English)0.507 oz.
Bottom ShapeConical
Closure IncludedNo
Diameter (Metric)17.5 mm
GraduatedYes
Product TypeCentrifuge Tube
Max. RCF1800 x g
Unit SizeCase