Orbital and Reciprocating Shaking Water Bath Accessories: Flask Trays
Actual product may vary
Boekel Scientific
Orbital and Reciprocating Shaking Water Bath Accessories: Flask Trays
For use with Boekel Scientific Orbital and Reciprocating Shaking Water Baths.
Have Questions?
Catalog NumberType
FIS05-450-102Universal Flask Tray
Catalog number FIS05-450-102
Price (SGD)
2,627.00
Each
Type:
Universal Flask Tray
Price (SGD)
2,627.00
Each
  • Hold variety of flask sizes.
  • Universal Flask Tray: Adjustable Flask Tray that switches mixing pattern by switching orientation.
  • Perforated Flask Tray: Perforated Diffuser Tray that enables Boekel Scientific Shaking Water Bath to be used as a non-shaking water bath.
Specifications
DescriptionUniversal Flask Tray for Boekel Scientific Shaking Water Bath Model 290400
For Use With (Equipment)Boekel Scientific Shaking Water Bath Model 290400
TypeUniversal Flask Tray
Holds43 x 25 mL, 26 x 50 mL, 15 x 100 mL, 11 x 250 mL, 6 x 500 mL, 3 x 100 mL Flasks
MaterialStainless Steel
Shipping Weight (English)5 lb.
Shipping Weight (Metric)2.2 kg
Unit SizeEach