Cimarec i Poly 15 and Multipoint Stirrers
Actual product may vary
Thermo Scientific
Cimarec i Poly 15 and Multipoint Stirrers
Conduct stirring applications with these stirrers, which are 100% maintenance- and wear-free, in a fully synchronized multipoint platform.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberModel
FIS04-979-203Poly 15
50093538Multipoint 15
FKS10145751Multipoint 15
50093557Multipoint 6
50094596Poly 15
Catalog number FIS04-979-203
Price (SGD)
6,663.00
Each
Model:
Poly 15
Price (SGD)
6,663.00
Each
Conduct stirring applications with Thermo Scientific™ Cimarec™ i Poly 15 and Multipoint Stirrers for reliable, safe performance. These stirrers are 100% maintenance- and wear-free, in a fully synchronized multipoint platform.
Features:
  • Large, flat, easy-to-clean work surfaces
  • Precise microprocessor control with bright digital display
  • Easily recall last used settings
  • Gradual start acceleration is gentle and ensures good magnetic coupling
  • Inductive drive ensures completely synchronized stirring at all positions
  • Operating conditions are -10°C to +40°C at 95%RH
Poly 15 Multipoint Stirrer:
  • Rugged, powder-coated, stainless-steel housing
  • Stir speeds up to 990rpm
  • Suitable for 15 × 250mL beakers or 6 × 1L flasks
  • IP32 protection rated
Advanced Multipoint 6/15 Stirrers:
  • Chemical-resistant, sealed stainless-steel housing
  • Adjustable power settings are separate from the speed control that decreases power consumption and lowers heat output
  • Smooth and even stirring as low as 80rpm
  • Stir speeds up to 2000rpm
  • Stir volumes up to 3L per multipoint
  • IP62 protection rated; easily cleaned with running water
Includes:
All models are 100-240V and contain a cord set with various plugs.
Recommended for:
Mass screening applications and other applications requiring multiple stirring points
Warranty: 5 years
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Speed130 to 990 rpm
Stirring Positions15
Top Plate MaterialStainless Steel
Capacity5 x 250 mL (Beaker), 6 x 1 L (Flask)
DisplayDigital
ShapeSquare
Voltage100/240 V
Wattage10 w
Dimensions (L x W x H)16.5 x 9.4 x 1.4 in. (420 x 240 x 35 mm)
Height (English)1.4 in.
Height (Metric)35 mm
Length (English)16.5 in.
Length (Metric)420 mm
Width (English)9.4 in.
Width (Metric)240 mm
Certifications/ComplianceCE
DescriptionPoly 15; Capacity 15 x 250mL beakers or 6 x 1L flasks; 130 to 990rpm; 10W
ModelPoly 15
Protection ClassIP32
Temperature Range (Metric)-10°C to +40°C (Operating)
TypeStirrer
Weight (English)13.2 lb.
Weight (Metric)6 kg
Unit SizeEach