Membrane Filters
Actual product may vary
BrandTech
Membrane Filters
PTFE membrane filters, conveniently packaged in easy-to-use blister packs.
Have Questions?
Catalog NumberPore SizeQuantity
FIS04-977-1580.2 μmEach
Catalog number FIS04-977-158
Price (SGD)
67.00
Pack
Pore Size:
0.2 μm
Quantity:
Each
Price (SGD)
67.00
Pack
Allows continued use after years of use. BrandTech™ BRAND™ macro™ Pipet Controller Accessories available for order include Suction Bellows, Valve System, Non-sterile 3μ Membrane Filters, and Silicone Adapters.
BRAND™ replacement filters for accu-jet™ and macro pipette controllers.
Features:
  • Hydrophobic PTFE membrane with durable polypropylene housing.
  • Available in 0.2 μm and 3.0 μm pore sizes.
  • The 0.2 μm filter is also available in a sterile version
  • Each filter is conveniently packaged in a blister pack for easy handling and storage. Available individually or in packs of 10.
  • accujet™ S and accujet™ pro are compatible with both the 0.2 μm and 3.0 μm filters.
  • macro pipette controller is compatible only with the 3.0 μm filters.
Specifications
For Use Withaccu-jet S
Pore Size0.2 μm
SterilitySterile
TypePipette Filter
Filter TypePTFE
For Use With (Application)Reduces cross-contamination, protects against over aspiration and allows pipetting to Class A tolerances
For Use With (Equipment)BRAND accu-jet Pipette Controller
QuantityEach
Unit SizePack