U.S. Standard Brass and Stainless Steel Test Sieve Receivers
U.S. Standard Brass and Stainless Steel Test Sieve Receivers
Actual product may vary
U.S. Standard Brass and Stainless Steel Test Sieve Receivers
Sieve reciever to suit all your testing, sampling and classification applications in the field, facility or lab.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (English) OuterMaterial
FIS04-888A12 in.Stainless Steel
FIS04-887B8 in.Stainless Steel
FIS04-886B8 in.Brass
Catalog number FIS04-888A
Price (SGD)
410.00
EA
Add to cart
Diameter (English) Outer:
12 in.
Material:
Stainless Steel
Price (SGD)
410.00
EA
Add to cart
Sieve reciever to suit all your testing, sampling and classification applications in the field, facility or lab.
Fit all 8 and 12 in. diameter (20.3 and 30.5cm) sieves.
Specifications
Depth (English)3 in.
Depth (Metric)7.6 cm
TypeFull Height Bottom Pan
Diameter (English) Outer12 in.
MaterialStainless Steel
For Use With (Equipment)Test Sieve
Height (English)4.25 in.
Height (Metric)10.7 cm
Diameter (Metric) Outer305 mm
Frame MaterialStainless Steel
Mesh MaterialStainless Steel
Unit SizeEA