Brass Test Sieves, 8 in. dia. x 2 in. D
Brass Test Sieves, 8 in. dia. x 2 in. D
Actual product may vary
Brass Test Sieves, 8 in. dia. x 2 in. D
Brass. Aperture sizes from 5.00 in. opening to 500 mesh.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSieve Number
FIS04-883H5/16
FIS04-881BB200
FIS04-881DD270
FIS04-881G10
FIS04-881J14
FIS04-881K16
FIS04-881L18
FIS04-881M20
FIS04-881P30
FIS04-881Q35
FIS04-881R40
FIS04-881U60
FIS04-881V70
FIS04-881X100
FIS04-881Z140
FIS04-883A1
FIS04-883D5/8
Catalog number FIS04-883H
Price (SGD)
244.00
EA
Add to cart
Sieve Number:
5/16
Price (SGD)
244.00
EA
Add to cart
Brass. Aperture sizes from 5.00 in. opening to 500 mesh.
  • All sieves are serialized.
  • The list below does not represent the complete ASTM E 11 specifications; other sizes are available.
Note:
A basic certificate of compliance comes with every test sieve. Additional certification is offered for customers who have more stringent QC programs; contact Thermo Fisher Customer Service.
Specifications
Depth (English)2 in.
Depth (Metric)5.08 cm
Pore Size8 mm
TypeFull Height Test Sieve
Diameter (English) Outer8 in.
MaterialBrass
Height (English)2.62 in.
IncludesTest sieve certificate
Height (Metric)6.65 cm
Tyler Standard2.5 Mesh
U.S. StandardNo. 4
For Use With (Application)Ideal for particle measurement in agricultural, bio industry, chemical, cosmetics, general industrial, fertilizer, foods, minerals, pharmaceuticals, Plastics and Metal products industries
Diameter (Metric) Outer203 mm
Frame MaterialBrass
Screen Size2.5 Mesh
Mesh MaterialStainless Steel
Sieve Number5/16
Unit SizeEA