Search Thermo Fisher Scientific
| Catalog Number | Pore Size |
|---|---|
| FIS04-881-83 | 63 μm |
| FIS04-881-11 | 4.75 mm |
| FIS04-881-21 | 425 μm |
| FIS04-881-28 | 45 μm |
Brass with Stainless Steel cloth. Aperture sizes from 5.00 in. opening to 500 mesh.
All sieves are serialized and come with a Test Sieve Certificate.
The list below does not represent the complete ASTM E 11 specifications; other sizes are available. Contact your Fisher representative for more information.