Conflikt™ Detergent Disinfectant
Actual product may vary
Fisherbrand
Conflikt™ Detergent Disinfectant
Prediluted ready-to-use spray quaternary ammonium disinfectant.
Have Questions?
Catalog NumberVolume (English)Container Type
FIS04-355-521 gal.Bottle
Catalog number FIS04-355-52
Price (SGD)
107.00
Each
Volume (English):
1 gal.
Container Type:
Bottle
Price (SGD)
107.00
Each
Effective against TB, HBV, and a wide range of viruses, bacteria, pathogenic fungi, mold, and mildew, including HIV-1, E. coli, and Canine Parvovirus.
  • Environmentally Friendly: Item contains environmental attributes or features (e.g. water conserving, lead-free, etc.) as described in the item detail
  • Recycled: Item contains recycled content as described in the item detail
  • Non-toxic: Item is non-toxic
Specifications
Product TypeDisinfectant
Color-codedNo
FormLiquid
pH11.7
ScentRegular Scent
SterilityNon-sterile
AppearanceClear to Light Yellow
Boiling Point200°F
Certifications/ComplianceEPA, OSHA, USDA
Composition0.11% n-Alkyl Dimethyl Benzyl Ammonium Chlorides, 0.11% n-Alkyl Dimethyl Ethylbenzyl Ammonium Chlo
Container TypeBottle
For Use With (Application)Cleans and disinfects hard surfaces that may harbor Viruses, Bacteria and Fungi
Size1 gal.
Volume (English)1 gal.
Volume (Metric)3.8 L
Unit SizeEach
Contents & Storage
50°F to 120°F