Conflikt™ Detergent Disinfectant
Actual product may vary
Conflikt™ Detergent Disinfectant
Prediluted ready-to-use spray quaternary ammonium disinfectant.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (English)Container Type
FIS04-355-3432 oz.Trigger Spray Bottle
FIS04-355-521 gal.Bottle
Catalog number FIS04-355-34
Price (SGD)
91.00
Each
Volume (English):
32 oz.
Container Type:
Trigger Spray Bottle
Price (SGD)
91.00
Each
Effective against TB, HBV, and a wide range of viruses, bacteria, pathogenic fungi, mold, and mildew, including HIV-1, E. coli, and Canine Parvovirus.
  • Environmentally Friendly: Item contains environmental attributes or features (e.g. water conserving, lead-free, etc.) as described in the item detail
  • Recycled: Item contains recycled content as described in the item detail
  • Non-toxic: Item is non-toxic
Specifications
Product TypeDisinfectant
Color-codedNo
FormLiquid
pH11.7
ScentRegular Scent
SterilityNon-sterile
AppearanceClear to Light Yellow
Boiling Point200°F
Certifications/ComplianceEPA, OSHA, USDA
Composition0.11% n-Alkyl Dimethyl Benzyl Ammonium Chlorides, 0.11% n-Alkyl Dimethyl Ethylbenzyl Ammonium Chlo
Container TypeTrigger Spray Bottle
For Use With (Application)Cleans and disinfects hard surfaces that may harbor Viruses, Bacteria and Fungi
Size32 oz.
Volume (English)32 oz.
Volume (Metric)946 mL
Unit SizeEach
Contents & Storage
50°F to 120°F