ELIMINase™ Decontaminant
Actual product may vary
ELIMINase™ Decontaminant
Eliminates RNase, DNase, and DNA from laboratory surfaces
Have Questions?
Catalog NumberVolume (Metric)
FIS04-355-32950 mL
Catalog number FIS04-355-32
Price (SGD)
116.00
Each
Volume (Metric):
950 mL
Price (SGD)
116.00
Each
  • Safe on glass, plastics, stainless steel, pipetters, gel boxes, and benchtops
  • Also acts as a radioactive decontaminant
  • Leaves no residue that could degrade RNA and DNA samples or interfere with subsequent enzymatic reaction
  • Works instantly
  • Environmentally Friendly: Item contains environmental attributes or features (e.g. water conserving, lead-free, etc.) as described in the item detail
  • Recycled: Item contains recycled content as described in the item detail
  • Non-toxic: Item is non-toxic
Specifications
Product TypeLaboratory Decontaminant
Volume (English)32 oz.
Volume (Metric)950 mL
For Use With (Application)Eliminates RNase, DNase and DNA Contamination
Unit SizeEach