Adjustable-Flame Burners
Adjustable-Flame Burners
Actual product may vary
Adjustable-Flame Burners
Ideal for very slow heating or quick boiling
Have Questions?
Catalog number FIS03-962Q
Price (SGD)
-
Ideal for very slow heating or quick boiling
  • Universal straight-tube burners have flames adjustable from 0.75 to 12 in. (20 to 305mm) Obtain heat output of over 1465w (5000 BTU/hr.)
  • Consume 5 cu. ft./hr. of gas
  • Flame retainer (supplied) prevents flame lifting or extinguishing
  • Adjustable air and gas controls ensure complete combustion
  • Tapered, serrated gas connection
  • Nonferrous, rustproof construction
  • Separate models for natural and LP gases
Specifications
Product TypeAdjustable Flame Burner
Gas TypeNatural Gas
Diameter (English)0.5 in.
Diameter (Metric)1.3 cm
For Use With (Application)Very Slow Heating or Quick Boiling
Heat Value800 to 1200 BTU
Height (English)6.5 in.
Height (Metric)16.5 cm
Unit SizeEA