accuFlame™ Natural Gas Burner
accuFlame™ Natural Gas Burner
Actual product may vary
accuFlame™ Natural Gas Burner
accuFlame™ Natural Gas Burner
accuFlame™ Natural Gas Burner
Stainless-steel grid top for better flame adjustment
Have Questions?
Catalog number FIS03-902Q
Price (SGD)
-
Stainless-steel grid top for better flame adjustment
  • Provides better Meker Cone definition for easier fine tuning
  • Distributes flame through 100 separate openings for uniform heating; flame arrester prevents flashback
  • Provides twice the heat output (2930w/10,000 BTU/hr.) of straight-tube burners
  • Features Venturi tube for thorough mixing of gas and air, and adjustable gas orifices and air shutter for complete combustion
  • Burns natural gas at 10 cu. ft./hr.
  • Rustproof chrome-plated brass body with no-tip, zinc-aluminum alloy base
Specifications
Product TypeNatural Gas Burner
Gas TypeNatural Gas
MaterialBrass, Stainless Steel, Zinc
Diameter (English)1.18 in.
Diameter (Metric)3 cm
For Use With (Application)Venturi Tube for Thorough Mixing of Air and Gas and Adjustable Air Shutter and Gas Orifices for Complete Combustion
Heat Value150 to 600 BTU
Height (English)7.4 in.
Height (Metric)19 cm
Unit SizeEA