Check Order Status
Glass Funnel
Actual product may vary
Glass Funnel
Provides for easy buret filling.
Have Questions?
Catalog Number
Diameter (Metric)Top
FIS03-865
40 mm
Catalog number
FIS03-865
Price (SGD)
20.00
EA
Add to cart
Diameter (Metric)Top:
40 mm
Price (SGD)
20.00
EA
Add to cart
Material: 33 expansion borosilicate glass
Stem length: 1.37 in. (35mm)
Outer diameter (top): 1.57 in. (40mm)
Overall height: 2.55 in. (65mm)
Autoclavable
Specifications
Diameter (Metric)Top
40 mm
Material
Borosilicate Glass
Height (Metric)
65 mm
Autoclavable
Autoclavable
Diameter (Metric) Plate
1.57 in.
Length (Metric) Stem
35mm
Product Type
Glass Funnel